Gene Transcripts

Gene Symbol
LY6H
Entrez Gene ID
4062
Gene Name
lymphocyte antigen 6 complex, locus H
Gene Aliases
NMLY6
Chromosome Location
Chr. 8: 143157914 - 143160636 on Build GRCh38
UniGene ID
Hs.159590
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130478.1 NP_001123950.1 4 320
NM_001135655.1 NP_001129127.1 3 300
NM_002347.4 NP_002338.3 3 264
XM_017013413.1 XP_016868902.1 2 278
XM_017013414.1 XP_016868903.1 3 325
GenBank mRNA AB012293.1 - 3 244
AK314139.1 - 4 320
BC028894.1 - 3 290 4939118
BC030192.1 - 3 224 5199071
BI914261.1 - 2 105
BP347868.1 - 3 228
CR457034.1 - 2 98
DB471677.1 - 3 264

View Full Product Details