Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000429.2 | NP_000420.1 | 6 | 845 | |
XM_005269842.4 | XP_005269899.1 | 7 | 1075 | ||
XM_005269843.4 | XP_005269900.1 | 6 | 964 | ||
GenBank mRNA | BC018359.1 | - | 6 | 812 | |
D49357.1 | - | 6 | 608 | ||
X69078.1 | - | 6 | 661 |