Gene Transcripts

Gene Symbol
MBD1
Entrez Gene ID
4152
Gene Name
methyl-CpG binding domain protein 1
Gene Aliases
CXXC3, PCM1, RFT
Chromosome Location
Chr. 18: 50266882 - 50281774 on Build GRCh38
UniGene ID
Hs.405610
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204136.1 NP_001191065.1 15 2200
NM_001204137.1 NP_001191066.1 16 2275
NM_001204138.1 NP_001191067.1 16 2272
NM_001204139.1 NP_001191068.1 15 2200
NM_001204140.1 NP_001191069.1 15 2107
NM_001204141.1 NP_001191070.1 14 2050
NM_001204143.1 NP_001191072.1 13 1675
NM_001204151.2 NP_001191080.1 13 1993
NM_001323942.1 NP_001310871.1 16 2275
NM_001323947.1 NP_001310876.1 16 2230
NM_001323949.1 NP_001310878.1 16 2267
NM_001323950.1 NP_001310879.1 15 2152
NM_001323951.1 NP_001310880.1 15 2155
NM_001323952.1 NP_001310881.1 14 1957
NM_001323953.1 NP_001310882.1 13 2000
NM_001323954.1 NP_001310883.1 13 1963
NM_002384.2 NP_002375.1 13 1894
NM_015844.2 NP_056669.2 14 2032
NM_015845.3 NP_056670.2 13 1993
NM_015846.3 NP_056671.2 15 2200
NM_015847.3 NP_056723.2 14 2053
XM_005258271.2 XP_005258328.1 15 1947
XM_006722456.2 XP_006722519.1 16 2022
XM_011525991.1 XP_011524293.1 15 1884
XM_011525993.2 XP_011524295.1 16 2022
XM_011525994.2 XP_011524296.1 16 2022
XM_011525998.2 XP_011524300.1 15 1944
XM_011525999.1 XP_011524301.1 14 1809
XM_011526001.1 XP_011524303.1 14 1779
XM_011526002.1 XP_011524304.1 15 1884
XM_011526003.2 XP_011524305.1 15 1884
XM_011526006.1 XP_011524308.1 13 1641
XM_011526007.1 XP_011524309.1 14 1809
XM_017025751.1 XP_016881240.1 16 1977
XM_017025752.1 XP_016881241.1 15 1902
XM_017025753.1 XP_016881242.1 15 1947
XM_017025754.1 XP_016881243.1 16 1977
XM_017025755.1 XP_016881244.1 15 1947
XM_017025756.1 XP_016881245.1 15 1884
XM_017025757.1 XP_016881246.1 16 1977
XM_017025758.1 XP_016881247.1 16 1977
XM_017025759.1 XP_016881248.1 16 1977
XM_017025760.1 XP_016881249.1 15 1947
XM_017025761.1 XP_016881250.1 15 1884
XM_017025762.1 XP_016881251.1 15 1902
XM_017025763.1 XP_016881252.1 15 1899
XM_017025764.1 XP_016881253.1 15 1884
XM_017025765.1 XP_016881254.1 14 1779
XM_017025766.1 XP_016881255.1 15 1854
XM_017025767.1 XP_016881256.1 15 1854
XM_017025768.1 XP_016881257.1 14 1779
XM_017025769.1 XP_016881258.1 14 1809
XM_017025770.1 XP_016881259.1 14 1779
XM_017025771.1 XP_016881260.1 14 1779
XM_017025772.1 XP_016881261.1 14 1734
XM_017025773.1 XP_016881262.1 13 1710
XM_017025774.1 XP_016881263.1 13 1710
XM_017025775.1 XP_016881264.1 13 1641
XM_017025776.1 XP_016881265.1 13 1641
XM_017025777.1 XP_016881266.1 14 1809
GenBank mRNA AB209765.1 - 10 1160
AF078830.1 - 14 1763
AF078831.1 - 12 1556
AF078832.1 - 14 1595
AF078833.1 - 12 1457
AK225889.1 - 15 2110
AK291519.1 - 15 2201
AK295401.1 - 16 2007
AK300759.1 - 13 1675
AK302004.1 - 16 1967
BC033242.1 - 14 1758 5432943
BC047353.1 - 14 1735
Y10746.1 - 14 1755

View Full Product Details