Gene Transcripts

Gene Symbol
MCM5
Entrez Gene ID
4174
Gene Name
minichromosome maintenance complex component 5
Gene Aliases
CDC46, P1-CDC46
Chromosome Location
Chr. 22: 35400068 - 35454942 on Build GRCh38
UniGene ID
Hs.517582
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006739.3 NP_006730.2 3 348
XM_006724242.3 XP_006724305.1 3 403
GenBank mRNA AK122853.1 - 3 325
AK130620.1 - 302
AK223323.1 - 3 338
AK291136.1 - 3 329
AK301121.1 - 338
AK308343.1 - 3 298
AK312533.1 - 3 328
BC000142.1 - 3 300 2900229
BC003656.2 - 2 274 3542201
BM787690.1 - 3 351
CR456517.1 - 3 288
D83986.1 - 3 336
X74795.2 - 2 276

View Full Product Details