Gene Transcripts
- Gene Symbol
- MYO7A
- Entrez Gene ID
- 4647
- Gene Name
- myosin VIIA
- Gene Aliases
- DFNA11, DFNB2, MYOVIIA, MYU7A, NSRD2, USH1B
- Chromosome Location
- Chr. 11: 77128192 - 77215241 on Build GRCh38
- UniGene ID
- Hs.370421
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000260.3 |
NP_000251.3
|
4, 5 |
550 |
|
NM_001127179.2 |
NP_001120651.2
|
4, 5 |
550 |
|
NM_001127180.1 |
NP_001120652.1
|
4, 5 |
544 |
|
XM_011545044.2 |
XP_011543346.1
|
4, 5 |
622 |
|
XM_011545046.2 |
XP_011543348.2
|
3, 4 |
383 |
|
XM_017017778.1 |
XP_016873267.1
|
3, 4 |
383 |
|
XM_017017779.1 |
XP_016873268.1
|
3, 4 |
383 |
|
XM_017017780.1 |
XP_016873269.1
|
3, 4 |
383 |
|
XM_017017781.1 |
XP_016873270.1
|
3, 4 |
383 |
|
XM_017017782.1 |
XP_016873271.1
|
3, 4 |
383 |
|
XM_017017783.1 |
XP_016873272.1
|
3, 4 |
383 |
|
XM_017017784.1 |
XP_016873273.1
|
3, 4 |
383 |
|
XM_017017785.1 |
XP_016873274.1
|
3, 4 |
383 |
|
XM_017017786.1 |
XP_016873275.1
|
3, 4 |
383 |
|
XM_017017787.1 |
XP_016873276.1
|
3, 4 |
383 |
|
XM_017017788.1 |
XP_016873277.1
|
3, 4 |
383 |
|
GenBank mRNA |
BC144519.1 |
- |
5, 6 |
592 |
|
U34227.1 |
- |
4, 5 |
545 |
|
U39226.1 |
- |
4, 5 |
550 |
|
U55208.1 |
- |
4, 5 |
545 |
|
U55209.1 |
- |
4, 5 |
545 |
|
View Full Product Details