Gene Transcripts

Gene Symbol
NPTX2
Entrez Gene ID
4885
Gene Name
neuronal pentraxin 2
Gene Aliases
NARP, NP-II, NP2
Chromosome Location
Chr. 7: 98617285 - 98629869 on Build GRCh38
UniGene ID
Hs.3281
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002523.2 NP_002514.1 2 617
GenBank mRNA BC009924.2 - 2 609
BC034781.1 - 2 605 5169124
BC035339.1 - 2 610 5198692
BC048275.1 - 2 575 5165672
BI561105.1 - 2 622
U26662.1 - 2 333

View Full Product Details