Gene Transcripts

Gene Symbol
PCK2
Entrez Gene ID
5106
Gene Name
phosphoenolpyruvate carboxykinase 2, mitochondrial
Gene Aliases
PEPCK, PEPCK-M, PEPCK2
Chromosome Location
Chr. 14: 24094131 - 24104132 on Build GRCh38
UniGene ID
Hs.75812
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018073.2 NP_001018083.2 2 366
NM_001308054.1 NP_001294983.1 2 627
NM_004563.3 NP_004554.3 2 366
XM_006720158.2 XP_006720221.1 2 359
GenBank mRNA AK129934.1 - 2 184
AK301400.1 - 2 210
AK307913.1 - 2 595
AK311710.1 - 2 597
BC001454.2 - 2 156 3138368
BQ957219.1 - 2 584
BX248272.1 - 2 140
CR456913.1 - 2 91
DB028389.1 - 2 359
HY022221.1 - 3 266
KU178243.1 - 2 91
KU178244.1 - 2 91
KU178245.1 - 2 91
X92720.1 - 2 157

View Full Product Details