Gene Transcripts
- Gene Symbol
- CERCAM
- Entrez Gene ID
- 51148
- Gene Name
- cerebral endothelial cell adhesion molecule
- Gene Aliases
- CEECAM1, GLT25D3
- Chromosome Location
- Chr. 9: 128405993 - 128437351 on Build GRCh38
- UniGene ID
- Hs.495230
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286760.1 |
NP_001273689.1
|
8, 9 |
986 |
|
NM_016174.4 |
NP_057258.3
|
8, 9 |
1462 |
|
XM_005252035.1 |
XP_005252092.1
|
10, 11 |
2372 |
|
XM_011518762.1 |
XP_011517064.1
|
11, 12 |
2529 |
|
XM_011518763.2 |
XP_011517065.1
|
8, 9 |
2080 |
|
XM_017014794.1 |
XP_016870283.1
|
12, 13 |
2271 |
|
GenBank mRNA |
AB040935.1 |
- |
9, 10 |
3975 |
|
AF218016.1 |
- |
3, 4 |
212 |
|
AK074519.1 |
- |
7, 8 |
898 |
|
AK074523.1 |
- |
7, 8 |
872 |
|
AK297929.1 |
- |
7, 8 |
958 |
|
AK307753.1 |
- |
5, 6 |
739 |
|
AK314294.1 |
- |
8, 9 |
1033 |
|
BC011811.2 |
- |
1, 2 |
105 |
|
BC098432.1 |
- |
8, 9 |
1060 |
6672104 |
BC108698.1 |
- |
8, 9 |
1073 |
|
BC119698.1 |
- |
8, 9 |
935 |
40116832 |
BC119699.1 |
- |
8, 9 |
935 |
40116834 |
View Full Product Details