Gene Transcripts
- Gene Symbol
- TMEM14C
- Entrez Gene ID
- 51522
- Gene Name
- transmembrane protein 14C
- Gene Aliases
- C6orf53, HSPC194, MSTP073, NET26, bA421M1.6
- Chromosome Location
- Chr. 6: 10722915 - 10731129 on Build GRCh38
- UniGene ID
- Hs.519557
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001165258.1 |
NP_001158730.1
|
4, 5 |
590 |
|
NM_016462.3 |
NP_057546.1
|
4, 5 |
576 |
|
GenBank mRNA |
AF151028.1 |
- |
4, 5 |
386 |
|
AF164962.1 |
- |
3, 4 |
321 |
|
AK223462.1 |
- |
4, 5 |
274 |
|
AK309591.1 |
- |
4, 5 |
324 |
|
AK310191.1 |
- |
4, 5 |
323 |
|
AL702149.1 |
- |
4, 5 |
410 |
|
BC002496.2 |
- |
4, 5 |
311 |
3050609 |
BC010086.2 |
- |
4, 5 |
340 |
2907072 |
CX784985.1 |
- |
4, 5 |
576 |
|
View Full Product Details