Gene Transcripts
- Gene Symbol
- DHRS7
- Entrez Gene ID
- 51635
- Gene Name
- dehydrogenase/reductase 7
- Gene Aliases
- CGI-86, SDR34C1, retDSR4, retSDR4
- Chromosome Location
- Chr. 14: 60144119 - 60169864 on Build GRCh38
- UniGene ID
- Hs.59719
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001322280.1 |
NP_001309209.1
|
6, 7 |
1008 |
|
NM_001322281.1 |
NP_001309210.1
|
6, 7 |
1164 |
|
NM_001322282.1 |
NP_001309211.1
|
5, 6 |
917 |
|
NM_016029.3 |
NP_057113.1
|
6, 7 |
1157 |
|
GenBank mRNA |
AF126782.1 |
- |
|
1103 |
|
AF151844.1 |
- |
6, 7 |
1050 |
|
AK313285.1 |
- |
6, 7 |
1082 |
|
AL117420.1 |
- |
6, 7 |
1091 |
|
AY359031.1 |
- |
6, 7 |
966 |
|
BC000637.2 |
- |
6, 7 |
993 |
3344010 |
BC007337.2 |
- |
6, 7 |
1011 |
3616802 |
View Full Product Details