Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278167.1 | NP_001265096.1 | 8 | 1062 | |
NM_001278169.1 | NP_001265098.1 | 8 | 830 | ||
NM_016082.4 | NP_057166.4 | 8 | 914 | ||
NM_016408.3 | NP_057492.2 | 8 | 1062 | ||
XM_011528856.2 | XP_011527158.1 | 5 | 1798 | ||
XM_011528857.2 | XP_011527159.1 | 7 | 674 | ||
XM_011528858.1 | XP_011527160.1 | 7 | 913 | ||
XM_011528859.2 | XP_011527161.1 | 7 | 673 | ||
XM_011528860.1 | XP_011527162.1 | 5 | 373 | ||
XM_017027876.1 | XP_016883365.1 | 7 | 812 | ||
XM_017027877.1 | XP_016883366.1 | 2 | 126 | ||
GenBank mRNA | AF152097.1 | - | 9 | 1040 | |
AF161516.1 | - | 8 | 1010 | ||
AK023992.1 | - | 8 | 1012 | ||
AK027449.1 | - | 8 | 862 | ||
AK222745.1 | - | 8 | 1012 | ||
AK292075.1 | - | 8 | 780 | ||
AK295342.1 | - | 8 | 998 | ||
AK314532.1 | - | 8 | 1012 | ||
AY462283.1 | - | 8 | 1013 | ||
AY462284.1 | - | 8 | 1013 | ||
BC050706.1 | - | 8 | 1005 | 4418659 | |
CX785345.1 | - | 3 | 269 | ||
DR156307.1 | - | 7 | 671 | ||
KU178645.1 | - | 7 | 900 |