Gene Transcripts
- Gene Symbol
- ANLN
- Entrez Gene ID
- 54443
- Gene Name
- anillin actin binding protein
- Gene Aliases
- FSGS8, Scraps, scra
- Chromosome Location
- Chr. 7: 36389803 - 36453791 on Build GRCh38
- UniGene ID
- Hs.62180
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001284301.2 |
NP_001271230.1
|
4, 5 |
1098 |
|
NM_001284302.2 |
NP_001271231.1
|
4, 5 |
1098 |
|
NM_018685.4 |
NP_061155.2
|
4, 5 |
1098 |
|
XM_006715746.1 |
XP_006715809.1
|
4, 5 |
1095 |
|
XM_006715747.3 |
XP_006715810.1
|
4, 5 |
1095 |
|
XM_017012354.1 |
XP_016867843.1
|
4, 5 |
1095 |
|
XM_017012355.1 |
XP_016867844.1
|
4, 5 |
1095 |
|
XM_017012356.1 |
XP_016867845.1
|
4, 5 |
1095 |
|
GenBank mRNA |
AK291254.1 |
- |
4, 5 |
1030 |
|
AK299803.1 |
- |
4, 5 |
800 |
|
BC070066.1 |
- |
4, 5 |
1080 |
4825802 |
BE885510.1 |
- |
|
325 |
|
View Full Product Details