Gene Transcripts

Gene Symbol
ODAM
Entrez Gene ID
54959
Gene Name
odontogenic, ameloblast asssociated
Gene Aliases
APIN
Chromosome Location
Chr. 4: 70195728 - 70204576 on Build GRCh38
UniGene ID
Hs.143811
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017855.3 NP_060325.3 4 229
XM_005265698.3 XP_005265755.1 5 293
XM_006714245.3 XP_006714308.1 5 270
XM_017008331.1 XP_016863820.1 3 185
GenBank mRNA AK098169.1 - 1 295
BG181363.1 - 3 187
BG194365.1 - 4 232
EF113908.1 - 4 231

View Full Product Details