Gene Transcripts

Gene Symbol
VPS37C
Entrez Gene ID
55048
Gene Name
VPS37C, ESCRT-I subunit
Gene Aliases
Chromosome Location
Chr. 11: 61130256 - 61161444 on Build GRCh38
UniGene ID
Hs.523715
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017966.4 NP_060436.4 3 259
XM_005274077.3 XP_005274134.1 3 300
GenBank mRNA AK097326.1 - 3 236
AK302384.1 - 3 327
AL834261.1 - 3 267
BC141827.1 - 3 222 40148285
BC142702.1 - 3 222 40148798
BM450171.1 - 2 229

View Full Product Details