Gene Transcripts

Gene Symbol
UBR7
Entrez Gene ID
55148
Gene Name
ubiquitin protein ligase E3 component n-recognin 7 (putative)
Gene Aliases
C14orf130
Chromosome Location
Chr. 14: 93207056 - 93229215 on Build GRCh38
UniGene ID
Hs.648806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175748.3 NP_786924.2 4 594
NR_038150.1 3 460
GenBank mRNA AK097477.1 - 4 579
AK315492.1 - 4 393
BC015046.1 - 4 393 3908032
BC017013.1 - 3 238 3883770
BC051819.1 - 4 356 6171393
BX248249.1 - 4 353
DA158479.1 - 3 234

View Full Product Details