Gene Transcripts
- Gene Symbol
- INTS10
- Entrez Gene ID
- 55174
- Gene Name
- integrator complex subunit 10
- Gene Aliases
- C8orf35, INT10
- Chromosome Location
- Chr. 8: 19817149 - 19852078 on Build GRCh38
- UniGene ID
- Hs.512627
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018142.2 |
NP_060612.2
|
1, 2 |
250 |
|
XM_005273555.3 |
XP_005273612.1
|
1, 2 |
508 |
|
XM_005273556.3 |
XP_005273613.1
|
1, 2 |
292 |
|
XM_005273557.3 |
XP_005273614.1
|
1, 2 |
292 |
|
XM_006716355.2 |
XP_006716418.1
|
1, 2 |
266 |
|
XM_017013606.1 |
XP_016869095.1
|
1, 2 |
292 |
|
XM_017013607.1 |
XP_016869096.1
|
1, 2 |
508 |
|
XM_017013608.1 |
XP_016869097.1
|
1, 2 |
73 |
|
XM_017013609.1 |
XP_016869098.1
|
1, 2 |
266 |
|
GenBank mRNA |
AK001431.1 |
- |
1, 2 |
171 |
|
BM547763.1 |
- |
1, 2 |
250 |
|
View Full Product Details