Gene Transcripts
- Gene Symbol
- PPP2R5D
- Entrez Gene ID
- 5528
- Gene Name
- protein phosphatase 2 regulatory subunit B'delta
- Gene Aliases
- B56D, MRD35
- Chromosome Location
- Chr. 6: 42984499 - 43012345 on Build GRCh38
- UniGene ID
- Hs.533308
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270476.1 |
NP_001257405.1
|
10, 11 |
1285 |
|
NM_006245.3 |
NP_006236.1
|
10, 11 |
1268 |
|
NM_180976.2 |
NP_851307.1
|
10, 11 |
1172 |
|
NM_180977.2 |
NP_851308.1
|
8, 9 |
950 |
|
GenBank mRNA |
AB000634.1 |
- |
10, 11 |
1103 |
|
AB000635.1 |
- |
8, 9 |
797 |
|
AB451342.1 |
- |
10, 11 |
1089 |
|
AB451357.1 |
- |
10, 11 |
1089 |
|
AK122701.1 |
- |
10, 11 |
1162 |
|
AK290604.1 |
- |
10, 11 |
1081 |
|
AK291797.1 |
- |
10, 11 |
1176 |
|
AK299696.1 |
- |
10, 11 |
1141 |
|
BC001095.2 |
- |
10, 11 |
1152 |
3507280 |
BC001175.1 |
- |
10, 11 |
1079 |
3355395 |
BC010692.1 |
- |
10, 11 |
1162 |
3853892 |
D78360.1 |
- |
10, 11 |
1070 |
|
L76702.1 |
- |
10, 11 |
1276 |
|
View Full Product Details