Gene Transcripts

Gene Symbol
ZCCHC8
Entrez Gene ID
55596
Gene Name
zinc finger CCHC-type containing 8
Gene Aliases
Chromosome Location
Chr. 12: 122471599 - 122501073 on Build GRCh38
UniGene ID
Hs.37706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017612.4 NP_060082.2 9, 10 1106
XM_006719491.2 XP_006719554.1 6, 7 791
XM_011538555.2 XP_011536857.1 8, 9 1648
XM_017019606.1 XP_016875095.1 9, 10 1026
GenBank mRNA AK027702.1 - 9, 10 975
AK074638.1 - 9, 10 1004
AK291174.1 - 5, 6 441
AL157433.1 - 7, 8 2515
BC017704.2 - 2, 3 230
BC065918.1 - 9, 10 945 5752053

View Full Product Details