Gene Transcripts

Gene Symbol
POMGNT1
Entrez Gene ID
55624
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
Gene Aliases
GNTI.2, GnT I.2, LGMD2O, MEB, MGAT1.2, gnT-I.2
Chromosome Location
Chr. 1: 46188681 - 46220305 on Build GRCh38
UniGene ID
Hs.525134
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243766.1 NP_001230695.1 18, 19 2263
NM_001290129.1 NP_001277058.1 17, 18 1684
NM_001290130.1 NP_001277059.1 17, 18 1650
NM_017739.3 NP_060209.3 18, 19 1776
XM_005271010.1 XP_005271067.1 18, 19 1771
XM_006710755.1 XP_006710818.1 18, 19 1736
XM_006710756.1 XP_006710819.1 18, 19 1771
XM_011541760.2 XP_011540062.1 17, 18 1628
XM_017001690.1 XP_016857179.1 18, 19 1736
GenBank mRNA AB057356.1 - 18, 19 1706
AF250859.2 - 15, 16 1365
AK000284.1 - 18, 19 1753
AK055301.1 - 6 2513
AK056186.1 - 18, 19 2263
AK225033.1 - 18, 19 1754
AK301923.1 - 17, 18 1650
AK302379.1 - 17, 18 1660
AK316399.1 - 17, 18 1657
AY358592.1 - 18, 19 1741
BC001471.2 - 18, 19 1770 3138971
CR749700.1 - 15, 16 2521

View Full Product Details