Gene Transcripts
- Gene Symbol
- ASF1B
- Entrez Gene ID
- 55723
- Gene Name
- anti-silencing function 1B histone chaperone
- Gene Aliases
- CIA-II
- Chromosome Location
- Chr. 19: 14119509 - 14136628 on Build GRCh38
- UniGene ID
- Hs.26516
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018154.2 |
NP_060624.1
|
1, 2 |
285 |
|
GenBank mRNA |
AB104486.1 |
- |
1, 2 |
113 |
|
AF279307.1 |
- |
1, 2 |
206 |
|
AK001288.1 |
- |
1, 2 |
244 |
|
AK001466.1 |
- |
1, 2 |
276 |
|
AK223080.1 |
- |
1, 2 |
239 |
|
AK302134.1 |
- |
1, 2 |
246 |
|
BC007726.2 |
- |
1, 2 |
209 |
4303318 |
BC010014.2 |
- |
1, 2 |
219 |
4304756 |
BC036521.1 |
- |
1, 2 |
249 |
5270216 |
CN426445.1 |
- |
1, 2 |
620 |
|
CR457235.1 |
- |
1, 2 |
113 |
|
View Full Product Details