Gene Transcripts

Gene Symbol
PRR11
Entrez Gene ID
55771
Gene Name
proline rich 11
Gene Aliases
Chromosome Location
Chr. 17: 59155499 - 59206709 on Build GRCh38
UniGene ID
Hs.631750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018304.3 NP_060774.2 3 491
GenBank mRNA AK001891.1 - 3 244
AK225377.1 - 3 244
AK225380.1 - 3 249
AL711011.1 - 3 258
BC008669.1 - 3 233 3867293
DA649803.1 - 3 491
DB106349.1 - 3 258
FJ161961.1 - 3 244

View Full Product Details