Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278945.1 | NP_001265874.1 | 2 | 428 | |
NM_018492.3 | NP_060962.2 | 2 | 486 | ||
XM_006716368.2 | XP_006716431.1 | 2 | 410 | ||
GenBank mRNA | AB027249.1 | - | 2 | 212 | |
AB027250.1 | - | 2 | 192 | ||
AF189722.1 | - | 2 | 176 | ||
AF237709.1 | - | 2 | 200 | ||
AK027291.1 | - | 2 | 191 | ||
AK301836.1 | - | 2 | 388 | ||
BC015191.1 | - | 2 | 200 | 4082846 | |
BI461026.1 | - | 2 | 229 | ||
DB028500.1 | - | 2 | 486 | ||
EU794685.1 | - | 2 | 223 |