Gene Transcripts

Gene Symbol
SMG9
Entrez Gene ID
56006
Gene Name
SMG9, nonsense mediated mRNA decay factor
Gene Aliases
C19orf61, F17127_1, HBMS
Chromosome Location
Chr. 19: 43727992 - 43754990 on Build GRCh38
UniGene ID
Hs.466875
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005259057.2 XP_005259114.1 6 1013
XM_011527113.1 XP_011525415.1 6 768
XM_011527114.1 XP_011525416.1 6 706
XM_011527115.1 XP_011525417.1 6 774
XM_011527116.1 XP_011525418.1 6 900
GenBank mRNA AK022948.1 - 6 985
AK309830.1 - 6 890
AL136606.1 - 6 939
BC008869.2 - 6 925 4131601

View Full Product Details