Gene Transcripts

Gene Symbol
FMN2
Entrez Gene ID
56776
Gene Name
formin 2
Gene Aliases
Chromosome Location
Chr. 1: 240013776 - 240475189 on Build GRCh38
UniGene ID
Hs.24889
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305424.1 NP_001292353.1 8, 9 4394
NM_020066.4 NP_064450.3 7, 8 4382
XM_017001837.1 XP_016857326.1 8, 9 4392
XM_017001839.1 XP_016857328.1 8, 9 3234
XM_017001840.1 XP_016857329.1 7, 8 2634
XM_017001841.1 XP_016857330.1 7, 8 3092
XM_017001842.1 XP_016857331.1 6, 7 2313
XM_017001843.1 XP_016857332.1 5, 6 2301
GenBank mRNA AB209153.1 - 8, 9 2987
AK297716.1 - 2, 3 152
AK297755.1 - 2, 3 398
DN990332.1 - 3, 4 386

View Full Product Details