Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253723.1 | NP_001240652.1 | 2 | 99 | |
NM_001253724.1 | NP_001240653.1 | 2 | 102 | ||
NM_020134.3 | NP_064519.2 | 2 | 182 | ||
GenBank mRNA | AF157634.1 | - | 1 | 24 | |
AF264015.1 | - | 2 | 186 | ||
AJ251275.1 | - | 1 | 24 | ||
BC002874.2 | - | 2 | 99 | 3940393 | |
BT006871.1 | - | 1 | 24 | ||
DA410946.1 | - | 2 | 102 |