Gene Transcripts

Gene Symbol
WRNIP1
Entrez Gene ID
56897
Gene Name
Werner helicase interacting protein 1
Gene Aliases
WHIP, bA420G6.2
Chromosome Location
Chr. 6: 2765341 - 2785745 on Build GRCh38
UniGene ID
Hs.236828
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020135.2 NP_064520.2 1 845
NM_130395.2 NP_569079.1 1 845
XM_005249232.3 XP_005249289.1 1 936
XM_017011060.1 XP_016866549.1 1 936
GenBank mRNA AB056152.1 - 1 846
AB209723.1 - 1 598
AF218313.1 - 1 232
AK094823.1 - 1 805
AK223593.1 - 1 779
AK315047.1 - 1 937
AK315471.1 - 1 814
BC018923.2 - 1 806 4109718

View Full Product Details