Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001128429.2 | NP_001121901.1 | 10 | 1640 | |
NM_001128430.1 | NP_001121902.1 | 10 | 1542 | ||
NM_001254949.1 | NP_001241878.1 | 2 | 462 | ||
NM_020159.4 | NP_064544.2 | 10 | 1640 | ||
NR_045644.1 | 11 | 1691 | |||
XM_017008463.1 | XP_016863952.1 | 10 | 1464 | ||
XM_017008464.1 | XP_016863953.1 | 10 | 1464 | ||
XM_017008465.1 | XP_016863954.1 | 10 | 1474 | ||
GenBank mRNA | AB032948.2 | - | 9 | 1428 | |
AK025302.1 | - | 5 | 680 | ||
AK027490.1 | - | 10 | 1538 | ||
AK301668.1 | - | 2 | 462 | ||
AL359929.1 | - | 1 | 70 | ||
AY008271.1 | - | 10 | 1443 | ||
BC045534.1 | - | 10 | 1595 | 5267669 |