Gene Transcripts
- Gene Symbol
- CIAPIN1
- Entrez Gene ID
- 57019
- Gene Name
- cytokine induced apoptosis inhibitor 1
- Gene Aliases
- Anamorsin, DRE2, PRO0915
- Chromosome Location
- Chr. 16: 57428169 - 57447528 on Build GRCh38
- UniGene ID
- Hs.4900
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308358.1 |
NP_001295287.1
|
2, 3 |
396 |
|
NM_020313.3 |
NP_064709.2
|
2, 3 |
396 |
|
GenBank mRNA |
AF248964.1 |
- |
2, 3 |
300 |
|
AK074263.1 |
- |
2, 3 |
237 |
|
AK295024.1 |
- |
1, 2 |
629 |
|
BC002568.2 |
- |
2, 3 |
238 |
3139226 |
BC024196.2 |
- |
2, 3 |
277 |
4816594 |
BC067303.1 |
- |
2, 3 |
396 |
30389410 |
BC071740.1 |
- |
2, 3 |
271 |
5563094 |
BG746807.1 |
- |
1 |
242 |
|
BJ991015.1 |
- |
2, 3 |
277 |
|
View Full Product Details