Gene Transcripts

Gene Symbol
SLC24A3
Entrez Gene ID
57419
Gene Name
solute carrier family 24 member 3
Gene Aliases
NCKX3
Chromosome Location
Chr. 20: 19212646 - 19722897 on Build GRCh38
UniGene ID
Hs.654790
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020689.3 NP_065740.2 9, 10 966
GenBank mRNA AF169257.2 - 9, 10 712
AF288087.1 - 9, 10 966

View Full Product Details