Gene Transcripts

Gene Symbol
MIER1
Entrez Gene ID
57708
Gene Name
MIER1 transcriptional regulator
Gene Aliases
ER1, MI-ER1
Chromosome Location
Chr. 1: 66924895 - 66988619 on Build GRCh38
UniGene ID
Hs.605432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077700.2 NP_001071168.2 4 377
NM_001077701.2 NP_001071169.1 3 233
NM_001077702.2 NP_001071170.2 5 451
NM_001077703.2 NP_001071171.2 4 377
NM_001077704.2 NP_001071172.1 3 233
NM_001146110.1 NP_001139582.1 5 454
NM_001146111.1 NP_001139583.1 5 451
NM_001146112.1 NP_001139584.1 5 412
NM_001278215.1 NP_001265144.1 5 451
NM_020948.3 NP_065999.2 5 451
XM_005271076.3 XP_005271133.1 5 489
XM_011541864.2 XP_011540166.1 6 396
XM_011541865.2 XP_011540167.1 6 357
XM_017001919.1 XP_016857408.1 6 373
XM_017001920.1 XP_016857409.1 6 390
XM_017001921.1 XP_016857410.1 6 418
XM_017001922.1 XP_016857411.1 6 393
XM_017001924.1 XP_016857413.1 5 345
XM_017001925.1 XP_016857414.1 5 450
XM_017001926.1 XP_016857415.1 6 389
XM_017001927.1 XP_016857416.1 4 262
XM_017001928.1 XP_016857417.1 4 338
XM_017001929.1 XP_016857418.1 5 319
XM_017001930.1 XP_016857419.1 4 263
XM_017001931.1 XP_016857420.1 5 297
GenBank mRNA AF515446.1 - 3 233
AF515447.1 - 4 314
AF515448.2 - 5 388
AK302061.1 - 4 377
AK307717.1 - 5 348
AL831987.2 - 5 393
AY124186.2 - 5 388
AY124187.1 - 4 314
AY124188.1 - 3 233
AY124189.2 - 5 388
AY124190.1 - 4 314
AY124191.1 - 3 233
AY124192.2 - 5 388
AY124193.1 - 4 314
AY124194.1 - 3 233
BC017423.1 - 3 210
BC066898.1 - 6 379
BC108726.1 - 5 332 5194296
CR627441.1 - 5 288

View Full Product Details