Gene Transcripts
- Gene Symbol
- PTPRH
- Entrez Gene ID
- 5794
- Gene Name
- protein tyrosine phosphatase, receptor type H
- Gene Aliases
- R-PTP-H, SAP1
- Chromosome Location
- Chr. 19: 55181247 - 55209506 on Build GRCh38
- UniGene ID
- Hs.179770
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001161440.2 |
NP_001154912.2
|
14, 15 |
2320 |
|
NM_002842.4 |
NP_002833.4
|
16, 17 |
2854 |
|
XM_011527183.1 |
XP_011525485.1
|
16, 17 |
2875 |
|
XM_011527188.1 |
XP_011525490.1
|
15, 16 |
2824 |
|
XM_017027056.1 |
XP_016882545.1
|
15, 16 |
2862 |
|
XM_017027057.1 |
XP_016882546.1
|
15, 16 |
2850 |
|
XM_017027058.1 |
XP_016882547.1
|
15, 16 |
2895 |
|
XM_017027059.1 |
XP_016882548.1
|
15, 16 |
2862 |
|
XM_017027060.1 |
XP_016882549.1
|
14, 15 |
2814 |
|
XM_017027061.1 |
XP_016882550.1
|
13, 14 |
2328 |
|
XM_017027062.1 |
XP_016882551.1
|
15, 16 |
2770 |
|
XM_017027063.1 |
XP_016882552.1
|
15, 16 |
2763 |
|
GenBank mRNA |
BC111715.1 |
- |
15, 16 |
2270 |
|
BC111716.1 |
- |
16, 17 |
2804 |
40008775 |
D15049.2 |
- |
16, 17 |
2828 |
|
View Full Product Details