Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022110.3 | NP_071393.2 | 2 | 448 | |
GenBank mRNA | AK027119.1 | - | 2 | 446 | |
AK291418.1 | - | 2 | 443 | ||
BC004168.2 | - | 2 | 433 | 2820874 | |
BC011966.1 | - | 2 | 448 | 3857383 | |
CR457374.1 | - | 1 | 178 | ||
KP861813.1 | - | 2 | 434 |