Gene Transcripts

Gene Symbol
TMEM91
Entrez Gene ID
641649
Gene Name
transmembrane protein 91
Gene Aliases
DSPC3, IFITMD6
Chromosome Location
Chr. 19: 41363966 - 41384083 on Build GRCh38
UniGene ID
Hs.709417
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042595.2 NP_001036060.1 3 348
NM_001098821.1 NP_001092291.1 3 610
NM_001098822.1 NP_001092292.1 3 610
NM_001098823.1 NP_001092293.1 3 610
NM_001098824.1 NP_001092294.1 3 610
NM_001098825.1 NP_001092295.1 3 418
GenBank mRNA AK130820.1 - 3 919
AL833095.1 - 3 580
BC063705.1 - 3 320 4804092
BF684543.1 - 3 410
BG697182.1 - 3 348
BM726200.1 - 3 385
BM931402.1 - 3 565
BX090160.1 - 2 293
R67551.1 - 2 295

View Full Product Details