Gene Transcripts

Gene Symbol
SPINK9
Entrez Gene ID
643394
Gene Name
serine peptidase inhibitor, Kazal type 9
Gene Aliases
LEKTI2
Chromosome Location
Chr. 5: 148323219 - 148341974 on Build GRCh38
UniGene ID
Hs.631798
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040433.1 NP_001035523.1 4 288
XM_017009709.1 XP_016865198.1 4 828
GenBank mRNA AY396740.1 - 4 288
BC137530.1 - 4 259 9021530
BC137531.1 - 4 259 9021531

View Full Product Details