Gene Transcripts

Gene Symbol
TMPPE
Entrez Gene ID
643853
Gene Name
transmembrane protein with metallophosphoesterase domain
Gene Aliases
Chromosome Location
Chr. 3: 33090416 - 33096801 on Build GRCh38
UniGene ID
Hs.443031
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039770.2 NP_001034859.2 2 391
GenBank mRNA AK126979.1 - 2 391
BC136872.1 - 1 317 9020863
BC136873.1 - 1 317 9020864

View Full Product Details