Gene Transcripts

Gene Symbol
RASSF10
Entrez Gene ID
644943
Gene Name
Ras association domain family member 10
Gene Aliases
Chromosome Location
Chr. 11: 13009423 - 13012106 on Build GRCh38
UniGene ID
Hs.134650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080521.2 NP_001073990.2 1 1415

View Full Product Details