Gene Transcripts

Gene Symbol
KLC2
Entrez Gene ID
64837
Gene Name
kinesin light chain 2
Gene Aliases
Chromosome Location
Chr. 11: 66257294 - 66267861 on Build GRCh38
UniGene ID
Hs.280792
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134774.1 NP_001128246.1 7 1001
NM_001134775.1 NP_001128247.1 8 1295
NM_001134776.1 NP_001128248.1 8 1174
NM_001318734.1 NP_001305663.1 8 1232
NM_022822.2 NP_073733.1 8 1269
XM_005274183.1 XP_005274240.1 8 1211
XM_005274184.1 XP_005274241.1 8 1232
XM_017018133.1 XP_016873622.1 6 1168
GenBank mRNA AK022449.1 - 8 1161
AK022907.1 - 8 1214
AK094593.1 - 6 871
AK126489.1 - 6 1653
AK315725.1 - 8 1164
AL136864.1 - 8 1195
BC034373.1 - 8 1157 4342879

View Full Product Details