Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023012.5 | NP_075388.2 | 8 | 1101 | |
NR_036434.1 | 9 | 1182 | |||
NR_036435.1 | 9 | 1201 | |||
NR_036436.1 | 8 | 1123 | |||
XM_005253601.1 | XP_005253658.1 | 8 | 1104 | ||
XM_005253602.2 | XP_005253659.1 | 8 | 1075 | ||
XM_005253604.1 | XP_005253661.1 | 9 | 1186 | ||
XM_011538688.1 | XP_011536990.1 | 8 | 1104 | ||
XM_011538689.1 | XP_011536991.1 | 11 | 1359 | ||
XM_017019830.1 | XP_016875319.1 | 8 | 1101 | ||
XM_017019831.1 | XP_016875320.1 | 9 | 1181 | ||
XM_017019832.1 | XP_016875321.1 | 11 | 1337 | ||
XM_017019833.1 | XP_016875322.1 | 9 | 1128 | ||
GenBank mRNA | AB212664.1 | - | 8 | 1087 | |
AF161432.1 | - | 4 | 579 | ||
AK001883.1 | - | 8 | 1061 | ||
AK023985.1 | - | 5 | 691 | ||
AK056114.1 | - | 9 | 1124 | ||
AK056256.1 | - | 8 | 1061 | ||
BC008684.1 | - | 8 | 1011 | 3853928 | |
BC067773.1 | - | 8 | 1071 | 5301415 | |
BX640711.1 | - | 8 | 1047 |