Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003192.2 | NP_003183.1 | 1 | 995 | |
GenBank mRNA | AK312681.1 | - | 1 | 937 | |
BC017479.1 | - | 2 | 1486 | 3357283 | |
BC020170.1 | - | 1 | 935 | 3536688 | |
BT007002.1 | - | 1 | 922 | ||
U61234.1 | - | 1 | 945 |