Gene Transcripts

Gene Symbol
LRRC3C
Entrez Gene ID
100505591
Gene Name
leucine rich repeat containing 3C
Gene Aliases
Chromosome Location
Chr. 17: 39927722 - 39944999 on Build GRCh38
UniGene ID
Hs.145136
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195545.1 NP_001182474.1 2 796
XM_011524158.2 XP_011522460.1 4 982
XM_017024003.1 XP_016879492.1 4 961

View Full Product Details