Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144981.2 | NP_001138453.1 | 6 | 587 | |
NM_001144982.2 | NP_001138454.1 | 5 | 507 | ||
NM_001144983.2 | NP_001138455.1 | 5 | 503 | ||
NM_001144984.2 | NP_001138456.1 | 4 | 466 | ||
NM_001144985.2 | NP_001138457.1 | 5 | 507 | ||
NM_001144986.2 | NP_001138458.1 | 5 | 503 | ||
NM_001198908.1 | NP_001185837.1 | 6 | 587 | ||
GenBank mRNA | AK308999.1 | - | 5 | 476 | |
BC037403.1 | - | 6 | 718 | ||
BC146975.1 | - | 6 | 481 | 9056903 | |
BC171867.1 | - | 5 | 398 | 9054521 | |
BC171872.1 | - | 5 | 401 | 9054526 | |
BC171874.1 | - | 4 | 360 | 9054528 | |
BC171880.1 | - | 5 | 397 | 9054534 | |
BC171881.1 | - | 4 | 360 | 9054535 | |
BC171882.1 | - | 5 | 397 | 9054536 | |
BC171884.1 | - | 5 | 401 | ||
DN999189.1 | - | 6 | 544 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001198910.1 | NP_001185839.1 | 4 | 466 | |
GenBank mRNA | AK301863.1 | - | 4 | 453 |