Gene Transcripts

Gene Symbol
USP1
Entrez Gene ID
7398
Gene Name
ubiquitin specific peptidase 1
Gene Aliases
UBP
Chromosome Location
Chr. 1: 62436304 - 62451804 on Build GRCh38
UniGene ID
Hs.35086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017415.1 NP_001017415.1 9 2108
NM_001017416.1 NP_001017416.1 9 2013
NM_003368.4 NP_003359.3 9 2538
GenBank mRNA AB014458.1 - 9 1889
AB208893.1 - 9 2169
AF117386.1 - 9 2025
AK096788.1 - 1 820
AK312423.1 - 9 2201
AL117503.1 - 1 77
AL117575.1 - 8 1860
BC023216.1 - 5 1373
BC050525.1 - 9 2242 6473568

View Full Product Details