Gene Transcripts

Gene Symbol
ZNF90
Entrez Gene ID
7643
Gene Name
zinc finger protein 90
Gene Aliases
HTF9
Chromosome Location
Chr. 19: 20077994 - 20127032 on Build GRCh38
UniGene ID
Hs.55304
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007138.1 NP_009069.1 1 54
GenBank mRNA AK298173.1 - 1 49
M61870.1 - 1 35

View Full Product Details