Gene Transcripts

Gene Symbol
GGCT
Entrez Gene ID
79017
Gene Name
gamma-glutamylcyclotransferase
Gene Aliases
C7orf24, CRF21, GCTG, GGC
Chromosome Location
Chr. 7: 30496621 - 30504841 on Build GRCh38
UniGene ID
Hs.530024
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199815.1 NP_001186744.1 1 204
NM_001199816.1 NP_001186745.1 1 204
NM_001199817.1 NP_001186746.1 1 204
NM_024051.3 NP_076956.1 1 204
NR_037669.1 1 204
GenBank mRNA AK315608.1 - 1 195
BC000625.2 - 1 199 3346822
BC005356.1 - 1 192 4102029
BC013937.1 - 1 165 3997629
BC019243.1 - 1 196 3828006
BE407390.1 - 1 133
BE885397.1 - 1 164
BG527593.1 - 1 193

View Full Product Details