Gene Transcripts
- Gene Symbol
- LENG1
- Entrez Gene ID
- 79165
- Gene Name
- leukocyte receptor cluster member 1
- Gene Aliases
- Chromosome Location
- Chr. 19: 54155639 - 54159743 on Build GRCh38
- UniGene ID
- Hs.590974
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF211966.1 |
- |
1 |
106 |
|
BC014986.1 |
- |
2 |
293 |
4877885 |
View Full Product Details