Gene Transcripts

Gene Symbol
CENPO
Entrez Gene ID
79172
Gene Name
centromere protein O
Gene Aliases
CENP-O, ICEN-36, MCM21R
Chromosome Location
Chr. 2: 24793115 - 24822376 on Build GRCh38
UniGene ID
Hs.731569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199803.2 NP_001186732.1 5 1046
NM_001322101.1 NP_001309030.1 6 1160
NM_024322.3 NP_077298.1 6 1179
NR_136182.1 5 964
NR_136183.1 5 1065
NR_136184.1 5 945
GenBank mRNA AK315483.1 - 6 805
AL514456.3 - 5 739
BC002870.1 - 6 836 3942844
BX406097.2 - 6 834
CX872478.1 - 5 582

View Full Product Details