Gene Transcripts

Gene Symbol
C19orf57
Entrez Gene ID
79173
Gene Name
chromosome 19 open reading frame 57
Gene Aliases
Chromosome Location
Chr. 19: 13882348 - 13906125 on Build GRCh38
UniGene ID
Hs.143288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024323.3 NP_077299.3 3 165
XM_006722885.3 XP_006722948.2 3 538
GenBank mRNA AK097431.1 - 3 165
BC119719.1 - 2 133 40117748

View Full Product Details