Gene Transcripts

Gene Symbol
C19orf57
Entrez Gene ID
79173
Gene Name
chromosome 19 open reading frame 57
Gene Aliases
Chromosome Location
Chr. 19: 13882348 - 13906125 on Build GRCh38
UniGene ID
Hs.143288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024323.3 NP_077299.3 8 1862
XM_006722885.3 XP_006722948.2 8 2235
XM_017027300.1 XP_016882789.1 8 2115
XM_017027301.1 XP_016882790.1 7 1886
GenBank mRNA AK097431.1 - 8 1862
BC002891.2 - 4 1081
BC012945.1 - 5 1531
BC119718.2 - 1 309
BC119719.1 - 7 1830 40117748

View Full Product Details