Gene Transcripts

Gene Symbol
PLEKHS1
Entrez Gene ID
79949
Gene Name
pleckstrin homology domain containing S1
Gene Aliases
C10orf81, HEL185, bA211N11.2
Chromosome Location
Chr. 10: 113751262 - 113782433 on Build GRCh38
UniGene ID
Hs.228320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024889.4 NP_079165.3 2, 3 353
XM_005270162.2 XP_005270219.1 2, 3 492
XM_005270163.2 XP_005270220.1 2, 3 492
XM_011540171.1 XP_011538473.1 2, 3 492
XM_011540172.1 XP_011538474.1 3, 4 260
XM_011540173.1 XP_011538475.1 2, 3 492
XM_011540174.2 XP_011538476.1 2, 3 492
XM_017016666.1 XP_016872155.1 2, 3 492
GenBank mRNA AK128811.1 - 2, 3 353

View Full Product Details