Gene Transcripts

Gene Symbol
FBXL18
Entrez Gene ID
80028
Gene Name
F-box and leucine rich repeat protein 18
Gene Aliases
Fbl18
Chromosome Location
Chr. 7: 5454448 - 5513813 on Build GRCh38
UniGene ID
Hs.623974
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024963.5 NP_079239.3 5 2226
GenBank mRNA BC004228.1 - 3 679
BC013435.1 - 3 828

View Full Product Details